Learn More
Biomatik Corporation Recombinant Human MOCO Sulphurase C-Terminal Domain Containing Protein 1 (MOSC1)
Supplier:  Biomatik Corporation RPU52375
Encompass_Preferred
Recombinant Human MOCO Sulphurase C-Terminal Domain Containing Protein 1 (MOSC1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: MOCO Sulphurase C-Terminal Domain Containing Protein 1 (MOSC1). Target Synonyms: Molybdenum Cofactor Sulphurase C-Terminal Domain Containing Protein; Mitochondrial amidoxime-reducing component 1. Accession Number: Q5VT66. Gene ID: 64757. Expression Region: Arg41~Leu335. Tag Info: N-terminal His Tag. Theoretical MW: 37kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0189