Learn More
Biomatik Corporation Recombinant Human Methyl CpG Binding Domain Protein 2 (MBD2)
Supplier:  Biomatik Corporation RPU52609
Encompass_Preferred
Recombinant Human Methyl CpG Binding Domain Protein 2 (MBD2) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Methyl CpG Binding Domain Protein 2 (MBD2). Target Synonyms: DMTase; NY-CO-41; Demethylase; Methyl-CpG-binding protein MBD2. Accession Number: Q9UBB5. Gene ID: 8932. Expression Region: Glu145~Ala411. Tag Info: N-terminal His Tag. Theoretical MW: 36kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0199