Learn More
Biomatik Corporation Recombinant Human Alpha-2-Heremans Schmid Glycoprotein (AHSG)
Supplier:  Biomatik Corporation RPU40186
Encompass_Preferred
Recombinant Human Alpha-2-Heremans Schmid Glycoprotein (AHSG) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Alpha-2-Heremans Schmid Glycoprotein (AHSG). Target Synonyms: aHSG; HSGa; A2HS; AHS; FETUA; HSG-A2; Fetuin-A; Alpha 2-HS Glycoprotein; Fetuin A; Alpha-2-Z-Globulin; Ba-alpha-2-Glycoprotein. Accession Number: P02765. Gene ID: 197. Expression Region: Ala19~Leu300. Tag Info: N-terminal His Tag. Theoretical MW: 42kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9872