Learn More
Biomatik Corporation Recombinant Mouse Pulmonary Surfactant Associated Protein A1 (SFTPA1)
Supplier:  Biomatik Corporation RPU53010
Encompass_Preferred
Recombinant Mouse Pulmonary Surfactant Associated Protein A1 (SFTPA1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Pulmonary Surfactant Associated Protein A1 (SFTPA1). Target Synonyms: PRL; SFTPA; PSAP; PSPA; SFTP; SPA; COLEC4; PSP-A; SFTP1; SFTPA1B; SP-A1; Collectin-4; Alveolar proteinosis protein; 35 kDa pulmonary surfactant-associated protein; Surfactant Associated Protein A. Accession Number: P35242. Expression Region: Asn21~Phe248. Tag Info: N-terminal His Tag. Theoretical MW: 30kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0215