Learn More
Biomatik Corporation Recombinant Human Interleukin 1 Theta (IL1q)
Supplier:  Biomatik Corporation RPU53467
Encompass_Preferred
Recombinant Human Interleukin 1 Theta (IL1q) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Interleukin 1 Theta (IL1q). Target Synonyms: IL1F10; IL38; FKSG75; IL-1HY2; IL-1F10; FIL1-Theta; Interleukin-38; Interleukin-1 Receptor Antagonist FKSG75; Interleukin 1 Family, Member 10. Accession Number: Q8WWZ1. Gene ID: 84639. Expression Region: Ala15~Trp152. Tag Info: N-terminal His Tag. Theoretical MW: 19kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0224