Learn More
Biomatik Corporation Recombinant Human Transient Receptor Potential Cation Channel Subfamily V, Member 1 (TRPV1)
Supplier:  Biomatik Corporation RPU54111
Encompass_Preferred
Recombinant Human Transient Receptor Potential Cation Channel Subfamily V, Member 1 (TRPV1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Transient Receptor Potential Cation Channel Subfamily V, Member 1 (TRPV1). Target Synonyms: VR1; OTRPC1; Vanilloid Receptor Subtype 1; Capsaicin receptor; Osm-9-like TRP channel 1; Vanilloid receptor 1. Accession Number: Q8NER1. Gene ID: 7442. Expression Region: Met1~Glu153. Tag Info: N-terminal His Tag. Theoretical MW: 24kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0237