Learn More
Biomatik Corporation Recombinant Human Thyroid Stimulating Hormone Receptor (TSHR)
Supplier:  Biomatik Corporation RPU55340
Encompass_Preferred
Recombinant Human Thyroid Stimulating Hormone Receptor (TSHR) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Thyroid Stimulating Hormone Receptor (TSHR). Target Synonyms: LGR3; HTSHR-I; Thyrotropin Receptor; TSH Receptor. Accession Number: P16473. Gene ID: 7253. Expression Region: Gly21~Gly413. Tag Info: N-terminal His Tag. Theoretical MW: 50kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0290