Learn More
Biomatik Corporation Recombinant Human Ferritin, Mitochondrial (FTMT)
Supplier:  Biomatik Corporation RPU55352
Encompass_Preferred
Recombinant Human Ferritin, Mitochondrial (FTMT) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Ferritin, Mitochondrial (FTMT). Target Synonyms: MtF. Accession Number: Q8N4E7. Gene ID: 94033. Expression Region: Leu49~Asn242. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0292