Learn More
Biomatik Corporation Human Interleukin 8 (IL8), Active Protein
Supplier:  Biomatik Corporation RPU55475
Encompass_Preferred
Human Interleukin 8 (IL8), Active Protein is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Interleukin 8 (IL8). Target Synonyms: CXCL8; AMCF-I; GCP1; K60; LECT; LUCT; LYNAP; MDNCF; MONAP; NAF; NAP1; SCYB8; TSG1; B-ENAP; Neutrophil-Activating Protein 1; Granulocyte Chemotactic Protein 1. Accession Number: p10145. Gene ID: 3576. Expression Region: Ser28~Ser99. Tag Info: N-terminal His Tag. Theoretical MW: 12kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0298