Learn More
Biomatik Corporation Rat Nerve Growth Factor (NGF), Active Protein
Supplier:  Biomatik Corporation RPU55488
Encompass_Preferred
Rat Nerve Growth Factor (NGF), Active Protein is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Nerve Growth Factor (NGF). Target Synonyms: NGFB; Beta-NGF; HSAN5; NGF-B; Beta-Nerve Growth Factor. Accession Number: P25427. Gene ID: 310738. Expression Region: Glu19~Gly241. Tag Info: N-terminal His Tag. Theoretical MW: 33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0302