Learn More
Biomatik Corporation Recombinant Glutathione S Transferase (GST)
Supplier:  Biomatik Corporation RPU56062
Encompass_Preferred
Recombinant Glutathione S Transferase (GST) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Universal. Target Name: Glutathione S Transferase (GST). Target Synonyms: GST-Tag. Accession Number: P08515. Expression Region: Met1~Lys218. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0324