Learn More
Biomatik Corporation Recombinant Human Cementum Protein 1 (CEMP1)
Supplier:  Biomatik Corporation RPU56246
Encompass_Preferred
Recombinant Human Cementum Protein 1 (CEMP1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cementum Protein 1 (CEMP1). Target Synonyms: CP-23; Cementum Protein-23; Cementoblastoma-derived protein 1. Accession Number: Q6PRD7. Gene ID: 752014. Expression Region: Met1~Gly247. Tag Info: N-terminal His Tag. Theoretical MW: 32kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0332