Learn More
Biomatik Corporation Human Peptidylglycine Alpha Amidating Monooxygenase (PAM), Active Protein
Supplier:  Biomatik Corporation RPU59232
Encompass_Preferred
Human Peptidylglycine Alpha Amidating Monooxygenase (PAM), Active Protein is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Peptidylglycine Alpha Amidating Monooxygenase (PAM). Target Synonyms: PAL; PHM; Peptidylamidoglycolate lyase; Peptidyl-Alpha-hydroxyglycine Alpha-amidating Lyase; Peptidylglycine Alpha-Hydroxylating Monooxygenase. Accession Number: P19021. Gene ID: 5066. Expression Region: Phe21~Cys288. Tag Info: N-terminal His Tag. Theoretical MW: 34kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0359