Learn More
Biomatik Corporation Recombinant Mouse Lipoprotein lipase (LPL)
Supplier:  Biomatik Corporation RPU59583
Encompass_Preferred
Recombinant Mouse Lipoprotein lipase (LPL) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Lipoprotein lipase (LPL). Target Synonyms: LIPD; Lipase, Lipoprotein. Accession Number: P11152. Gene ID: 16956. Expression Region: Ala28~Gly474. Tag Info: N-terminal His Tag. Theoretical MW: 54kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0364