Learn More
Biomatik Corporation Recombinant Human TNF receptor-associated factor 6 (TRAF6)
Supplier:  Biomatik Corporation RPC20097
Encompass_Preferred
Recombinant Human TNF receptor-associated factor 6 (TRAF6) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: TRAF6. Target Synonyms: E3 ubiquitin-protein ligase TRAF6Interleukin-1 signal transducerRING finger protein 85. Accession Number: Q9Y4K3. Expression Region: 1~522aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 75.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0373