Learn More
Biomatik Corporation Recombinant Mouse Bone Morphogenetic Protein 1 (BMP1)
Supplier:  Biomatik Corporation RPU40480
Encompass_Preferred
Recombinant Mouse Bone Morphogenetic Protein 1 (BMP1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Bone Morphogenetic Protein 1 (BMP1). Target Synonyms: PCOLC; PCP; TLD; mTld; Procollagen C Proteinase; Mammalian tolloid protein. Accession Number: P98063. Gene ID: 12153. Expression Region: Gly856~Lys982. Tag Info: N-terminal His Tag. Theoretical MW: 22/23kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9887