Learn More
Biomatik Corporation Recombinant Saccharomyces cerevisiae Nicotinamidase (PNC1)
Supplier:  Biomatik Corporation RPC20366
Encompass_Preferred
Recombinant Saccharomyces cerevisiae Nicotinamidase (PNC1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast). Target Name: PNC1. Target Synonyms: Nicotine deamidaseNAMase. Accession Number: P53184. Expression Region: 1~216aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 29kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0405