Learn More
Biomatik Corporation Recombinant Mouse High affinity immunoglobulin epsilon receptor subunit alpha (Fcer1a)
Supplier:  Biomatik Corporation RPC20732
Encompass_Preferred
Recombinant Mouse High affinity immunoglobulin epsilon receptor subunit alpha (Fcer1a) is a purified CF Transmembrane Protein. Purity: >90% as determined by SDS-PAGE. Host: In Vitro E. coli Expression System. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Fcer1a. Target Synonyms: Fc-epsilon RI-alpha. Accession Number: P20489. Expression Region: 24~250aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged. Theoretical MW: 44.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0423