Learn More
Biomatik Corporation Recombinant Human Annexin A2 (ANXA2)
Supplier:  Biomatik Corporation RPC20900
Encompass_Preferred
Recombinant Human Annexin A2 (ANXA2) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ANXA2. Target Synonyms: Annexin II; Annexin-2; Calpactin I heavy chain; Calpactin-1 heavy chain; Chromobindin-8; Lipocortin II; Placental anticoagulant protein IV; PAP-IVProtein I; p36. Accession Number: P07355. Expression Region: 2~339aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 54.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0431