Learn More
Biomatik Corporation Recombinant Human Brain-derived neurotrophic factor (BDNF)
Supplier:  Biomatik Corporation RPC20975
Encompass_Preferred
Recombinant Human Brain-derived neurotrophic factor (BDNF) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: BDNF. Target Synonyms: Abrineurin. Accession Number: P23560. Expression Region: 129~247aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 29.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0437