Learn More
Biomatik Corporation Recombinant Human Angiopoietin 1 (ANGPT1)
Supplier:  Biomatik Corporation RPU40248
Encompass_Preferred
Recombinant Human Angiopoietin 1 (ANGPT1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Angiopoietin 1 (ANGPT1). Target Synonyms: ANG1; AGP1; AGPT; ANG-I. Accession Number: Q15389. Gene ID: 284. Expression Region: Arg284~Cys452. Tag Info: N-terminal His Tag. Theoretical MW: 19kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9877