Learn More
Biomatik Corporation Recombinant Rabbit CD40 ligand (CD40LG), partial
Supplier:  Biomatik Corporation RPC21056
Encompass_Preferred
Recombinant Rabbit CD40 ligand (CD40LG), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rabbit (Oryctolagus cuniculus). Target Name: CD40LG. Target Synonyms: Tumor necrosis factor ligand superfamily member 5. Accession Number: G1SKP7. Expression Region: 113~261aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 20.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0446