Learn More
Biomatik Corporation Recombinant Human C-type lectin domain family 18 member A (CLEC18A)
Supplier:  Biomatik Corporation RPC21103
Encompass_Preferred
Recombinant Human C-type lectin domain family 18 member A (CLEC18A) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CLEC18A. Target Synonyms: Mannose receptor-like protein 2. Accession Number: A5D8T8. Expression Region: 27~446aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 62.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0452