Learn More
Biomatik Corporation Recombinant Human Cholesterol side-chain cleavage enzyme, mitochondrial (CYP11A1)
Supplier:  Biomatik Corporation RPC21177
Encompass_Preferred
Recombinant Human Cholesterol side-chain cleavage enzyme, mitochondrial (CYP11A1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CYP11A1. Target Synonyms: CYPXIA1Cholesterol desmolaseCytochrome P450 11A1Cytochrome P450(scc). Accession Number: P05108. Expression Region: 40~521aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 72.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0465