Learn More
Biomatik Corporation Recombinant Human ATP-dependent RNA helicase DDX3X (DDX3X)
Supplier:  Biomatik Corporation RPC21199
Encompass_Preferred
Recombinant Human ATP-dependent RNA helicase DDX3X (DDX3X) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: DDX3X. Target Synonyms: DEAD box protein 3, X-chromosomalDEAD box, X isoformHelicase-like protein 2. Accession Number: O00571. Expression Region: 2~662aa. Tag Info: N-Terminal 6Xhis-B2M-Tagged. Theoretical MW: 87.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0467