Learn More
Biomatik Corporation Recombinant Human 11-beta-hydroxysteroid dehydrogenase 1 (HSD11B1), partial
Supplier:  Biomatik Corporation RPC21473
Encompass_Preferred
Recombinant Human 11-beta-hydroxysteroid dehydrogenase 1 (HSD11B1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: HSD11B1. Target Synonyms: 11-beta-hydroxysteroid dehydrogenase 1; 11-DH; 11-beta-HSD1Short chain dehydrogenase/reductase family 26C member 1. Accession Number: P28845. Expression Region: 25~292aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 45.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0492