Learn More
Biomatik Corporation Recombinant Human Carcinoembryonic Antigen (CEA)
Supplier:  Biomatik Corporation RPU40630
Encompass_Preferred
Recombinant Human Carcinoembryonic Antigen (CEA) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Carcinoembryonic Antigen (CEA). Target Synonyms: CD66E; CD66; CEACAM5; Carcinoembryonic Antigen-related Cell Adhesion Molecule 5; Carcinoembryonic Antigen. Accession Number: P06731. Gene ID: 1048. Expression Region: Ala566~Gly698. Tag Info: N-terminal His Tag. Theoretical MW: 16-25kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9894