Learn More
Biomatik Corporation Recombinant Human Mannan-binding lectin serine protease 1 (MASP1)
Supplier:  Biomatik Corporation RPC21653
Encompass_Preferred
Recombinant Human Mannan-binding lectin serine protease 1 (MASP1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: MASP1. Target Synonyms: Complement factor MASP-3 Complement-activating component of Ra-reactive factor Mannose-binding lectin-associated serine protease 1MASP-1 Mannose-binding protein-associated serine protease Ra-reactive factor serine protease p100RaRF Serine protease 5. Accession Number: P48740. Expression Region: 20~699aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 93kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0511