Learn More
Biomatik Corporation Recombinant Mouse Collagen Type VI Alpha 1 (COL6a1)
Supplier:  Biomatik Corporation RPU40905
Encompass_Preferred
Recombinant Mouse Collagen Type VI Alpha 1 (COL6a1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Collagen Type VI Alpha 1 (COL6a1). Target Synonyms: COL6-A1; OPLL; Collagen Alpha-1(VI)chain. Accession Number: Q04857. Gene ID: 12833. Expression Region: Asn82~Phe212. Tag Info: N-terminal His Tag. Theoretical MW: 16kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9903