Learn More
Biomatik Corporation Recombinant Human Integrin Alpha 2 (ITGa2)
Supplier:  Biomatik Corporation RPU50414
Encompass_Preferred
Recombinant Human Integrin Alpha 2 (ITGa2) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Integrin Alpha 2 (ITGa2). Target Synonyms: CD49b; BR; GPIa; VLA-2; VLAA2; Platelet membrane glycoprotein Ia; Alpha 2 Subunit Of VLA-2 Receptor; Very Late Activation Protein 2 Receptor; Collagen receptor; VLA-2 alpha. Accession Number: P17301. Gene ID: 3673. Expression Region: Pro170~Ile361. Tag Info: N-terminal His Tag. Theoretical MW: 28kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0100