Learn More
Biomatik Corporation Recombinant Human Serine/Arginine Rich Splicing Factor 1 (SRSF1)
Supplier:  Biomatik Corporation RPU50451
Encompass_Preferred
Recombinant Human Serine/Arginine Rich Splicing Factor 1 (SRSF1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Serine/Arginine Rich Splicing Factor 1 (SRSF1). Target Synonyms: ASF; SF2; SFRS1; Splicing Factor 2; Splicing Factor,Arginine/Serine-Rich 1; Alternate Splicing Factor; Alternative Splicing Factor; pre-mRNA-splicing factor SF2, P33 subunit. Accession Number: Q07955. Gene ID: 6426. Expression Region: Cys16~Arg224. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0103