Learn More
Biomatik Corporation BSA Conjugated Progesterone (PG)
Supplier:  Biomatik Corporation RPU50785
Encompass_Preferred
BSA Conjugated Progesterone (PG) is a purified Conjugated Small Molecules. Purity: >95% by SDS-PAGE. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Universal. Target Name: Progesterone (PG). Target Synonyms: P4; Pregn-4-Ene-3,20-Dione. Tag Info: N-terminal His Tag. Theoretical MW: 314.5g/mol. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0113