Learn More
Biomatik Corporation Recombinant Human Platelet Derived Growth Factor BB (PDGF BB)
Supplier:  Biomatik Corporation RPU50975
Encompass_Preferred
Recombinant Human Platelet Derived Growth Factor BB (PDGF BB) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Platelet Derived Growth Factor BB (PDGF BB). Target Synonyms: PDGF-BB. Accession Number: P01127. Gene ID: 5155. Expression Region: Ser82~Thr190. Tag Info: N-terminal His Tag. Theoretical MW: 43/21kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0118