Learn More
Biomatik Corporation Recombinant Mouse Enolase, Neuron Specific (NSE)
Supplier:  Biomatik Corporation RPU51141
Encompass_Preferred
Recombinant Mouse Enolase, Neuron Specific (NSE) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Enolase, Neuron Specific (NSE). Target Synonyms: ENO2; Enolase 2; Gamma Enolase; 2-phospho-D-glycerate hydro-lyase; Neural enolase. Accession Number: P17183. Gene ID: 13807. Expression Region: Ser2~Leu434. Tag Info: N-terminal His Tag. Theoretical MW: 48kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0128