Learn More
Biomatik Corporation Recombinant Mouse Complement Component 3 (C3)
Supplier:  Biomatik Corporation RPU51145
Encompass_Preferred
Recombinant Mouse Complement Component 3 (C3) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Complement Component 3 (C3). Target Synonyms: ASP; CPAMD1; Complement Protein C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a anaphylatoxin; Acylation stimulating protein. Accession Number: P01027. Gene ID: 12266. Expression Region: Ala965~Arg1303. Tag Info: N-terminal His Tag. Theoretical MW: 42kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0130