Learn More
Biomatik Corporation Recombinant Human Proteasome Maturation Protein (POMP)
Supplier:  Biomatik Corporation RPU51291
Encompass_Preferred
Recombinant Human Proteasome Maturation Protein (POMP) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Proteasome Maturation Protein (POMP). Target Synonyms: PNAS-110; UMP1; Proteassemblin; Protein UMP1 homolog; Voltage-gated K channel beta subunit 4.1. Accession Number: Q9Y244. Gene ID: 51371. Expression Region: Met1~Leu141. Tag Info: N-terminal His Tag. Theoretical MW: 20kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0136