Learn More
Biomatik Corporation Recombinant Human Osteopontin (OPN)
Supplier:  Biomatik Corporation RPU51355
Encompass_Preferred
Recombinant Human Osteopontin (OPN) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Osteopontin (OPN). Target Synonyms: SPP1; BNSP; BSPI; ETA1; Secreted Phosphoprotein 1; Bone Sialoprotein I; Early T-Lymphocyte Activation 1; Nephropontin; Urinary stone protein; Uropontin. Accession Number: P10451. Gene ID: 6696. Expression Region: Ile17~Asn287. Tag Info: N-terminal His and GST Tag. Theoretical MW: 70kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0141