Learn More
Biomatik Corporation Recombinant Human Collagen Type IV Alpha 5 (COL4a5)
Supplier:  Biomatik Corporation RPU40901
Encompass_Preferred
Recombinant Human Collagen Type IV Alpha 5 (COL4a5) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Collagen Type IV Alpha 5 (COL4a5). Target Synonyms: COL4-A5; ASLN; ATS; CA54; Collagen Alpha-5(IV)chain; Alport Syndrome. Accession Number: P29400. Gene ID: 1287. Expression Region: Gly1461~Thr1685. Tag Info: N-terminal His Tag. Theoretical MW: 31kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9902