Learn More
Biomatik Corporation Recombinant Human Actinin Alpha 4 (ACTN4)
Supplier:  Biomatik Corporation RPU51550
Encompass_Preferred
Recombinant Human Actinin Alpha 4 (ACTN4) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Actinin Alpha 4 (ACTN4). Target Synonyms: FSGS; FSGS1; F-actin cross-linking protein; Non-muscle alpha-actinin 4. Accession Number: O43707. Gene ID: 81. Expression Region: Met1~Ser269. Tag Info: N-terminal His Tag. Theoretical MW: 38kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0149