Learn More
Biomatik Corporation Recombinant Human Proenkephalin (PENK)
Supplier:  Biomatik Corporation RPU51848
Encompass_Preferred
Recombinant Human Proenkephalin (PENK) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Proenkephalin (PENK). Target Synonyms: Preproenkephalin; Proenkephalin-A; Synenkephalin; Met-enkephalin; Met-enkephalin-Arg-Gly-Leu; Leu-enkephalin; Met-enkephalin-Arg-Phe. Accession Number: P01210. Gene ID: 5179. Expression Region: Glu25~Phe267. Tag Info: N-terminal His Tag. Theoretical MW: 35kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0166