Learn More
Biomatik Corporation Recombinant Mouse Aspartylglucosaminidase (AGA)
Supplier:  Biomatik Corporation RPU51949
Encompass_Preferred
Recombinant Mouse Aspartylglucosaminidase (AGA) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Aspartylglucosaminidase (AGA). Target Synonyms: AGU; ASRG; GA; N(4)-(beta-N-acetylglucosaminyl)-L-asparaginase; Glycosylasparaginase; N4-(N-acetyl-beta-glucosaminyl)-L-asparagine amidase. Accession Number: Q64191. Gene ID: 11593. Expression Region: Ser24~Gly260. Tag Info: N-terminal His Tag. Theoretical MW: 29kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0172