Learn More
Biomatik Corporation Recombinant Human Semaphorin 3E (SEMA3E)
Supplier:  Biomatik Corporation RPU52356
Encompass_Preferred
Recombinant Human Semaphorin 3E (SEMA3E) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Semaphorin 3E (SEMA3E). Target Synonyms: SEMAH; M-SemaK; M-sema H; coll-5; Sema Domain,Immunoglobulin Domain(Ig),Transmembrane Domain(TM)and Short Cytoplasmic Domain 3E. Accession Number: O15041. Gene ID: 9723. Expression Region: Arg32~Phe516. Tag Info: N-terminal His Tag. Theoretical MW: 59kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0186