Learn More
Biomatik Corporation Recombinant Human Crystallin Alpha B (CRYaB)
Supplier:  Biomatik Corporation RPU52465
Encompass_Preferred
Recombinant Human Crystallin Alpha B (CRYaB) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Crystallin Alpha B (CRYaB). Target Synonyms: CRY-AB; CRYA-B; CRYA2; CTPP2; HSPB5; Heat shock protein beta-5; Renal carcinoma antigen NY-REN-27; Rosenthal fiber component. Accession Number: P02511. Gene ID: 1410. Expression Region: Met1~Lys175. Tag Info: N-terminal His Tag. Theoretical MW: 24kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0191