Learn More
Biomatik Corporation Recombinant Mouse Sestrin 2 (SESN2)
Supplier:  Biomatik Corporation RPU53116
Encompass_Preferred
Recombinant Mouse Sestrin 2 (SESN2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Sestrin 2 (SESN2). Target Synonyms: HI95; SES2; SEST2. Accession Number: P58043. Gene ID: 230784. Expression Region: Ser41~Thr480. Tag Info: N-terminal His Tag. Theoretical MW: 54kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0217