Learn More
Biomatik Corporation Recombinant Human Purinergic Receptor P2X, Ligand Gated Ion Channel 7 (P2RX7)
Supplier:  Biomatik Corporation RPU53204
Encompass_Preferred
Recombinant Human Purinergic Receptor P2X, Ligand Gated Ion Channel 7 (P2RX7) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Purinergic Receptor P2X, Ligand Gated Ion Channel 7 (P2RX7). Target Synonyms: P2X7; P2X Purinoceptor 7; ATP receptor; Purinergic receptor. Accession Number: Q99572. Gene ID: 5027. Expression Region: Ser47~Val334. Tag Info: N-terminal His Tag. Theoretical MW: 28/32/35kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0219