Learn More
Biomatik Corporation Recombinant Human RAD51 Homolog (RAD51)
Supplier:  Biomatik Corporation RPU53302
Encompass_Preferred
Recombinant Human RAD51 Homolog (RAD51) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: RAD51 Homolog (RAD51). Target Synonyms: RAD51A; RECA; HsRad51; BRCC5; BRCA1/BRCA2-Containing Complex Subunit 5; DNA repair protein RAD51 homolog 1. Accession Number: Q06609. Gene ID: 5888. Expression Region: Ala2~Asp339. Tag Info: N-terminal His Tag. Theoretical MW: 41kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0221