Learn More
Biomatik Corporation Recombinant Bovine Alpha-Lactalbumin (aLA)
Supplier:  Biomatik Corporation RPU53514
Encompass_Preferred
Recombinant Bovine Alpha-Lactalbumin (aLA) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Bovine (Bos taurus). Target Name: Alpha-Lactalbumin (aLA). Target Synonyms: LALBA; LYZL7; Lactose Synthase B Protein; Lysozyme Like Protein 7. Accession Number: P00711. Gene ID: 281894. Expression Region: Glu20~Leu142. Tag Info: N-terminal His Tag. Theoretical MW: 16kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0227