Learn More
Biomatik Corporation Recombinant Human Plectin (PLEC)
Supplier:  Biomatik Corporation RPU53846
Encompass_Preferred
Recombinant Human Plectin (PLEC) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Plectin (PLEC). Target Synonyms: HD1; PCN; EBS1; EBSO; PLEC1b; PLTN; Hemidesmosomal protein 1; Intermediate Filament Binding Protein 500kDa; Epidermolysis Bullosa Simplex 1(Ogna). Accession Number: Q15149. Gene ID: 5339. Expression Region: Asp175~Pro400. Tag Info: N-terminal His Tag. Theoretical MW: 32kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0234