Learn More
Biomatik Corporation Recombinant Mouse Ubiquitin Cross Reactive Protein (UCRP)
Supplier:  Biomatik Corporation RPU53888
Encompass_Preferred
Recombinant Mouse Ubiquitin Cross Reactive Protein (UCRP) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Ubiquitin Cross Reactive Protein (UCRP). Target Synonyms: G1P2; IFI15; ISG15; Interferon Alpha-Inducible Protein clone IFI-15K; Interferon Induced Protein 15; Interferon-induced 17 kDa protein. Accession Number: Q64339. Gene ID: 100038882. Expression Region: Ala2~Ala161. Tag Info: N-terminal His Tag. Theoretical MW: 22kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0236