Learn More
Biomatik Corporation Recombinant Bovine Thrombospondin 1 (THBS1)
Supplier:  Biomatik Corporation RPU54163
Encompass_Preferred
Recombinant Bovine Thrombospondin 1 (THBS1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Bovine (Bos taurus). Target Name: Thrombospondin 1 (THBS1). Target Synonyms: TSP1; Thrombospondin-1p180. Accession Number: Q28178. Gene ID: 281530. Expression Region: Asn65~Cys270. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0238